6-O-Methyl-2',4''-bis-O-(trimethylsilyl)erythromycin9-[O-(1-ethoxy-1-methylethyl)oxime] ≥95%

Supplier: Matrix Scientific

076223-250MG 076223-1G
102545-066EA 684.62 USD
102545-066 102545-068
6-O-Methyl-2',4''-bis-O-(trimethylsilyl)erythromycin9-[O-(1-ethoxy-1-methylethyl)oxime] ≥95%
6-O-Methyl-2',4''-bis-O-(trimethylsilyl)erythromycin9-[O-(1-ethoxy-1-methylethyl)oxime]

Hazard: Irritant

Formula: C₄₈H₉₄N₂O₁₄Si₂
MDL Number: MFCD09263748
CAS Number: 119699-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR