Ethyl glyoxylate 50% in toluene , 95%
Supplier: Matrix Scientific
076220-100G
076220-25G
102545-054EA
76.92
USD
102545-054
102664-090
Ethyl glyoxylate 50% in toluene , 95%
Ethyl glyoxylate
Hazard: Flammable/Air Sensitive
Formula:
C₄H₆O₃ MW: 102.09 g/mol Boiling Pt: 110 °C Density: 1.003 Flash Pt: 7 °C |
MDL Number:
MFCD00044009 CAS Number: 924-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...