Methyl-4-amino-2-fluorobenzoate ≥95%

Supplier: Matrix Scientific

076217-1G 076217-5G 076217-10G
102545-036EA 33.85 USD
102545-036 102545-038 102545-040
Methyl-4-amino-2-fluorobenzoate ≥95%
Methyl 4-amino-2-fluorobenzoate

Hazard: Irritant

Formula: C₈H₈FNO₂
MDL Number: MFCD09037917
CAS Number: 73792-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR