5-Bromopyrimidine-2-carboxamide ≥95%

Supplier: Matrix Scientific

076216-250MG 076216-1G
102545-032EA 640 USD
102545-032 102545-034
5-Bromopyrimidine-2-carboxamide ≥95%
5-Bromopyrimidine-2-carboxamide

Hazard: Irritant

Formula: C₅H₄BrN₃O
MDL Number: MFCD00507192
CAS Number: 38275-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR