4-Chlorophthalic anhydride ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloroisobenzofuran-1,3-dione

076213-10G 076213-100G
102545-016EA 40 USD
102545-016 102545-018
4-Chlorophthalic anhydride ≥95%
4-Chlorophthalic anhydride

Hazard: Irritant

Formula: C₈H₃ClO₃
MW: 182.56 g/mol
MDL Number: MFCD00152354
CAS Number: 118-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR