2,5-Dimethoxyphenol ≥95%

Supplier: Matrix Scientific

076212-250MG 076212-1G 076212-5G
102545-010EA 456.92 USD
102545-010 102545-012 102545-014
2,5-Dimethoxyphenol ≥95%
2,5-Dimethoxyphenol

Hazard: Irritant

Formula: C₈H₁₀O₃
MDL Number: MFCD00094237
CAS Number: 18113-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR