N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Erlotinib Hydrochloride

076211-1G 076211-5G
102545-006EA 33.85 USD
102545-006 102545-008
N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride ≥95%
N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride

Hazard: Irritant

Formula: C₂₂H₂₄ClN₃O₄
MW: 429.901 g/mol
MDL Number: MFCD07781272
CAS Number: 183319-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR