(2,3-Dihydrobenzo[b][1,4]dioxin-5-yl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

076210-250MG 076210-1G 076210-5G
102545-000EA 340 USD
102545-000 102545-002 102545-004
(2,3-Dihydrobenzo[b][1,4]dioxin-5-yl)methanamine hydrochloride ≥95%
(2,3-Dihydrobenzo[b][1,4]dioxin-5-yl)methanamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClNO₂
CAS Number: 870851-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR