(5-Aminopyridin-3-yl)boronic acid hydrochloride ≥95%

Supplier: Matrix Scientific

076209-250MG 076209-1G
102544-996EA 1395.38 USD
102544-996 102544-998
(5-Aminopyridin-3-yl)boronic acid hydrochloride ≥95%
(5-Aminopyridin-3-yl)boronic acid hydrochloride

Hazard: Irritant

Formula: C₅H₈BClN₂O₂
MDL Number: MFCD12198141
CAS Number: 1310384-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR