(S)-Pyrrolidine-3-carboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

076205-1G 076205-5G
102544-978EA 710.77 USD
102544-978 102544-980
(S)-Pyrrolidine-3-carboxylic acid hydrochloride ≥95%
(S)-Pyrrolidine-3-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClNO₂
MDL Number: MFCD09025426
CAS Number: 1124369-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR