1-Bromo-4-fluorobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromofluorobenzene

076202-10G 076202-100G
102544-964EA 364.62 USD
102544-964 102544-966
1-Bromo-4-fluorobenzene ≥95%
1-Bromo-4-fluorobenzene

Hazard: Flammable

Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 150 °C
Density: 1.593
Flash Pt: 53 °C
MDL Number: MFCD00000342
CAS Number: 460-00-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR