2-Bromobenzylamine ≥95%

Supplier: Matrix Scientific

Synonyms: (2-Bromophenyl)methanamine

076201-1G 076201-5G 076201-10G
102544-958EA 47.08 USD
102544-958 102544-960 102544-962
2-Bromobenzylamine ≥95%
2-Bromobenzylamine

Hazard: Corrosive/Air Sensitive

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 118…119 °C (9 mmHg)
Melting Pt: 36 °C
MDL Number: MFCD00025572
CAS Number: 3959-05-5
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR