Xanthine ≥95%

Supplier: Matrix Scientific

Synonyms: 1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine

076195-10G 076195-100G
102544-932EA 44.62 USD
102544-932 102544-934
Xanthine ≥95%
Xanthine

Hazard: Irritant

Formula: C₅H₄N₄O₂
MW: 152.11 g/mol
MDL Number: MFCD00078453
CAS Number: 69-89-6
Merck Index: 12,10193

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR