7-Chloro-3,4-dihydroquinolin-2(1H)-one ≥95%

Supplier: Matrix Scientific

076194-1G 076194-5G
102544-928EA 640 USD
102544-928 102544-930
7-Chloro-3,4-dihydroquinolin-2(1H)-one ≥95%
7-Chloro-3,4-dihydroquinolin-2(1H)-one

Hazard: Irritant

Formula: C₉H₈ClNO
MW: 181.62 g/mol
MDL Number: MFCD09033162
CAS Number: 14548-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR