1-Iodo-3-(trifluoromethoxy)benzene ≥95%

Supplier: Matrix Scientific

076191-5G 076191-25G
102544-916EA 92.31 USD
102544-916 102544-918
1-Iodo-3-(trifluoromethoxy)benzene ≥95%
1-Iodo-3-(trifluoromethoxy)benzene

Hazard: Irritant

Formula: C₇H₄F₃IO
MW: 288.01 g/mol
MDL Number: MFCD01090992
CAS Number: 198206-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR