1-Amino-2-naphthol hydrochloride ≥95%

Supplier: Matrix Scientific

076190-1G 076190-5G 076190-10G
102544-910EA 61.54 USD
102544-910 102544-912 102544-914
1-Amino-2-naphthol hydrochloride ≥95%
1-Amino-2-naphthol hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₀ClNO
MW: 195.65 g/mol
MDL Number: MFCD00012561
CAS Number: 1198-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR