4-Hydroxyphthalic acid ≥95%

Supplier: Matrix Scientific

076189-5G 076189-25G
102544-906EA 364.62 USD
102544-906 102833-912
4-Hydroxyphthalic acid ≥95%
4-Hydroxyphthalic acid

Hazard: Irritant

Formula: C₈H₆O₅
MW: 182.13 g/mol
Melting Pt: ∼ 190 °C (dec.)
MDL Number: MFCD00013984
CAS Number: 610-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR