5-Methoxytryptamine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(5-Methoxy-1H-indol-3-yl)ethanamine, O-Methylserotonin hydrochloride
076187-5G
076187-100G
076187-25G
102544-892EA
72.31
USD
102544-892
102544-896
102552-022
5-Methoxytryptamine hydrochloride ≥95%
5-Methoxytryptamine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅ClN₂O Melting Pt: 245…250 °C |
MDL Number:
MFCD00005662 CAS Number: 66-83-1 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...