2-Propylmalonic acid ≥95%

Supplier: Matrix Scientific

076186-1G 076186-5G 076186-10G
102544-886EA 364.62 USD
102544-886 102544-888 102544-890
2-Propylmalonic acid ≥95%
2-Propylmalonic acid

Hazard: Irritant

Formula: C₆H₁₀O₄
MDL Number: MFCD03840631
CAS Number: 616-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR