Isopropylmalonic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Isopropylmalonic acid

076185-1G 076185-5G 076185-10G
102544-880EA 72.31 USD
102544-880 102544-882 102544-884
Isopropylmalonic acid ≥95%
Isopropylmalonic acid

Hazard: Irritant

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
MDL Number: MFCD00021717
CAS Number: 601-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR