3'-Chloro-2,2,2-trifluoroacetophenone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(3-Chlorophenyl)-2,2,2-trifluoroethanone
076183-1G
076183-5G
102544-872EA
213.85
USD
102544-872
102544-874
3'-Chloro-2,2,2-trifluoroacetophenone ≥95%
3'-Chloro-2,2,2-trifluoroacetophenone
Hazard: Irritant
Formula:
C₈H₄ClF₃O MW: 208.5650 g/mol |
MDL Number:
MFCD00461896 CAS Number: 321-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...