Luteolin ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4-benzopyrone, Natural Yellow 2, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one, 3',4',5,7-Tetrahydroxyflavone
076178-1G
076178-5G
076178-10G
102544-850EA
38.46
USD
102544-850
102544-852
102544-854
Luteolin ≥95%
Luteolin
Hazard: Irritant
Formula:
C₁₅H₁₀O₆ MW: 286.24 g/mol |
MDL Number:
MFCD00017309 CAS Number: 491-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...