4-Iodo-3-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

076176-1G 076176-5G 076176-10G
102544-838EA 143.08 USD
102544-838 102544-840 102544-842
4-Iodo-3-nitrobenzoic acid ≥95%
4-Iodo-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₄INO₄
MW: 293.02 g/mol
MDL Number: MFCD00502399
CAS Number: 35674-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR