5-Chloro-2-nitro-p-toluidine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-4-methyl-2-nitroaniline

076174-1G 076174-5G 076174-10G
102544-828EA 90.77 USD
102544-828 102544-830 102544-832
5-Chloro-2-nitro-p-toluidine ≥95%
5-Chloro-2-nitro-p-toluidine

Hazard: Irritant

Formula: C₇H₇ClN₂O₂
MW: 186.60 g/mol
MDL Number: MFCD00034066
CAS Number: 7149-80-6
UN: 2237
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR