2-(Quinolin-6-yl)acetic acid ≥95%

Supplier: Matrix Scientific

076173-1G 076173-5G
102544-824EA 187.69 USD
102544-824 102544-826
2-(Quinolin-6-yl)acetic acid ≥95%
2-(Quinolin-6-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₉NO₂
MW: 187.2 g/mol
MDL Number: MFCD07776794
CAS Number: 5622-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR