Fosinopril sodium ≥95%
Supplier: Matrix Scientific
Synonyms:
SQ 28,555, Sodium-(2S,4S)-4-cyclohexyl-1-(2-((2-methyl-1-(propionyloxy)propoxy)-(4-phenylbutyl)phosphoryl)acetyl)pyrrolidine-2-carboxylate, (2S,4S)-4-cyclohexyl-1-[2-[(2-methyl-1-propanoyloxypropoxy)-(4-phenylbutyl)phosphoryl]acetyl]pyrrolidine-2-carboxylate sodium, Sodium (2S,4S)-4-cyclohexyl-1-(2-((R)-((S)-2-methyl-1-(propionyloxy)propoxy)(4-phenylbutyl)phosphoryl)acetyl)pyrrolidine-2-carboxylate
076172-250MG
076172-1G
102544-820EA
427.69
USD
102544-820
102544-822
Fosinopril sodium ≥95%
Fosinopril sodium
Purity: 95+%
CAS: 88889-14-9
MDL Number: MFCD00865785
Molecular Formula: C30H45NNaO7P
Molecular Weight: 585,66
Hazard: Irritant
Formula:
C₃₀H₄₅NNaO₇P MW: 585.64 g/mol |
MDL Number:
MFCD23704184 CAS Number: 88889-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...