6-Methoxy-5-methylpyridin-3-amine hydrochloride ≥95%

Supplier: Matrix Scientific

076168-1G 076168-5G
102544-798EA 640 USD
102544-798 102544-800
6-Methoxy-5-methylpyridin-3-amine hydrochloride ≥95%
6-Methoxy-5-methylpyridin-3-amine hydrochloride

Hazard: Irritant

Formula: C₇H₁₁ClN₂O
MDL Number: MFCD05662388
CAS Number: 1159811-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR