3,5-Dimethoxybenzonitrile ≥95%

Supplier: Matrix Scientific

076167-5G 076167-10G 076167-100G
102544-792EA 69.23 USD
102544-792 102544-794 102544-796
3,5-Dimethoxybenzonitrile ≥95%
3,5-Dimethoxybenzonitrile

Hazard: Irritant

Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00001804
CAS Number: 19179-31-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR