4-Methyl-2-quinolinol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Lepidinol, 4-Methylquinolin-2-ol, 2-Hydroxy-4-methylquinoline
076165-1G
076165-5G
102544-784EA
569.23
USD
102544-784
102544-786
4-Methyl-2-quinolinol ≥95%
4-Methyl-2-quinolinol
Hazard: Irritant
Formula:
C₁₀H₉NO MW: 159.185 g/mol |
MDL Number:
MFCD00006745 CAS Number: 607-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...