2',3',5'-Tri-O-acetyluridine ≥95%

Supplier: Matrix Scientific

Synonyms: (2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3,4-diyl diacetate, Uridine triacetate, Uridine 2',3',5'-triacetate

076162-5G 076162-10G 076162-100G
102544-768EA 76.92 USD
102544-768 102544-770 102544-772
2',3',5'-Tri-O-acetyluridine ≥95%
2',3',5'-Tri-O-acetyluridine

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₉
MDL Number: MFCD00023795
CAS Number: 4105-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR