3,4-Dichloro-L-phenylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: H-Phe(3,4-DiCl)-OH, H-3,4-Dichloro-Phe-OH

076161-1G 076161-5G 076161-10G
102544-762EA 21.54 USD
102544-762 102544-764 102544-766
3,4-Dichloro-L-phenylalanine ≥95%
3,4-Dichloro-L-phenylalanine

Hazard: Irritant

Formula: C₉H₉Cl₂NO₂
MW: 234.076 g/mol
MDL Number: MFCD00191203
CAS Number: 52794-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR