2-Hydroxy-p-toluic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-4-methylbenzoic acid, m-Homosalicyclic acid, m-Cresotic acid, 4-Methylsalicylic acid

076159-10G 076159-100G
102544-754EA 36.92 USD
102544-754 102544-756
2-Hydroxy-p-toluic acid ≥95%
2-Hydroxy-p-toluic acid

Hazard: Irritant

Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00002454
CAS Number: 50-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR