Cytidine-5'-monophosphate ≥95%

Supplier: Matrix Scientific

076158-10G 076158-100G
102544-750EA 107.69 USD
102544-750 102544-752
Cytidine-5'-monophosphate ≥95%
Cytidine-5'-monophosphate

Hazard: Irritant

Formula: C₉H₁₄N₃O₈P
MW: 323.20 g/mol
MDL Number: MFCD00006544
CAS Number: 63-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR