ω-Bromo-4-methylacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: α-Bromo-4-methylacetophenone, Bromomethyl p-methylphenyl ketone, 2-Bromo-4'-methylacetophenone, 2-Bromo-1-(p-tolyl)ethanone, 4-Methylphenacyl bromide

076157-10G 076157-100G
102544-746EA 178.46 USD
102544-746 102544-748
ω-Bromo-4-methylacetophenone ≥95%
ω-Bromo-4-methylacetophenone

Hazard: Irritant

Formula: C₉H₉BrO
MW: 213.07 g/mol
MDL Number: MFCD00000203
CAS Number: 619-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR