3-Iodothiophene ≥95%

Supplier: Matrix Scientific

076153-1G 076153-5G
102544-730EA 20 USD
102544-730 102544-732
3-Iodothiophene ≥95%
3-Iodothiophene

Hazard: Irritant/Light Sensitive

Formula: C₄H₃IS
MW: 210.04 g/mol
Boiling Pt: 77 °C (11 mmHg)
Density: 2.06
Flash Pt: 78 °C
MDL Number: MFCD00037901
CAS Number: 10486-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR