1,8-Dihydroxynaphthalene ≥95%

Supplier: Matrix Scientific

076152-1G 076152-5G
102544-726EA 38.46 USD
102544-726 102544-728
1,8-Dihydroxynaphthalene ≥95%
1,8-Dihydroxynaphthalene

Hazard: Irritant/Store in Freezer

Formula: C₁₀H₈O₂
MW: 160.17
MDL Number: MFCD00042701
CAS Number: 569-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR