2-Phenylphenol sodium salt tetrahydrate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxybiphenyl Sodium Salt, Sodium 2-Phenylphenolate, Sodium 2-Hydroxybiphenyl, 2-Biphenylol Sodium Salt, Sodium o-Phenylphenate, Sodium 2-Biphenylolate

076151-10G 076151-100G
102544-722EA 36.92 USD
102544-722 102544-724
2-Phenylphenol sodium salt tetrahydrate ≥95%
2-Phenylphenol sodium salt tetrahydrate

Hazard: Irritant

Formula: C₁₂H₁₀O
MW: 192.19 g/mol
MDL Number: MFCD00002209
CAS Number: 6152-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR