DL-Cysteine hydrochloride ≥95%
Supplier: Matrix Scientific
076148-5G
076148-10G
076148-100G
102544-708EA
72.31
USD
102544-708
102544-710
102544-712
DL-Cysteine hydrochloride ≥95%
DL-Cysteine hydrochloride
Hazard: Irritant
Formula:
C₃H₇NO₂S·HCl |
MDL Number:
MFCD00064552 CAS Number: 10318-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...