L(+)-Serine ≥95%

Supplier: Matrix Scientific

Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

076146-10G 076146-100G
102544-700EA 15.38 USD
102544-700 102544-702
L(+)-Serine ≥95%
L(+)-Serine

Hazard: Irritant

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 259
MDL Number: MFCD00064224
CAS Number: 56-45-1
Merck Index: 14,00846

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR