Levulinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Oxovaleric acid, 4-Ketovaleric acid, 4-Oxopentanoic acid

076145-10G 076145-100G
102544-696EA 36.92 USD
102544-696 102544-698
Levulinic acid ≥95%
Levulinic acid

Hazard: Irritant

Formula: C₅H₈O₃
MW: 116.12 g/mol
MDL Number: MFCD00002796
CAS Number: 123-76-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR