Cyclohexanecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Hexahydrobenzoic acid

076144-100G 076144-25G
102544-694EA 29.23 USD
102544-694 102664-086
Cyclohexanecarboxylic acid ≥95%
Cyclohexanecarboxylic acid

Hazard: Irritant

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 231-233°C
Melting Pt: 29-32°C
Density: 1.048
Flash Pt: 118°C
MDL Number: MFCD00001461
CAS Number: 98-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR