5-Methylisophthalic acid ≥95%
Supplier: Matrix Scientific
076143-5G
076143-100G
102544-686EA
16.92
USD
102544-686
102544-690
5-Methylisophthalic acid ≥95%
5-Methylisophthalic acid
Hazard: Irritant
Formula:
C₉H₈O₄ Melting Pt: 296-300°C |
MDL Number:
MFCD00013986 CAS Number: 499-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...