1,4-Bis(diphenylphosphino)butane ≥95%

Supplier: Matrix Scientific

Synonyms: DPPB, 1,4-Bis(diphenylphosphino)butane, Butylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)butane, Butane-1,3-diylbis(diphenylphosphine)

076142-100G 076142-25G
102544-684EA 135.38 USD
102544-684 102833-904
1,4-Bis(diphenylphosphino)butane ≥95%
1,4-Bis(diphenylphosphino)butane

Hazard: Irritant

Formula: C₂₈H₂₈P₂
MW: 426.48 g/mol
Melting Pt: 132-136°C
MDL Number: MFCD00003051
CAS Number: 7688-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR