4-Chloro-4'-hydroxybenzophenone ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Chlorophenyl)(4-hydroxyphenyl)methanone

076140-10G 076140-100G
102544-670EA 46.15 USD
102544-670 102544-672
4-Chloro-4'-hydroxybenzophenone ≥95%
4-Chloro-4'-hydroxybenzophenone

Hazard: Irritant

Formula: C₁₃H₉ClO₂
MDL Number: MFCD00002357
CAS Number: 42019-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR