Suberic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1,8-Octanedioic acid, Octanedioic acid, 1,6-Hexanedicarboxylic acid

076139-100G 076139-500G
102544-668EA 78.46 USD
102544-668 102833-902
Suberic acid ≥95%
Suberic acid

Hazard: Irritant

Formula: C₈H₁₄O₄
MW: 174.2 g/mol
MDL Number: MFCD00004428
CAS Number: 505-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR