3,6-Dibromocarbazole ≥95%
Supplier: Matrix Scientific
Synonyms:
3,6-Dibromo-9H-carbazole
076137-100G
076137-25G
102544-660EA
135.38
USD
102544-660
102552-018
3,6-Dibromocarbazole ≥95%
3,6-Dibromocarbazole
Hazard: Irritant
Formula:
C₁₂H₇Br₂N MW: 325 g/mol |
MDL Number:
MFCD00004961 CAS Number: 6825-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...