Ethyl-3-hydroxybutyrate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-hydroxybutanoate, DL-Ethyl-3-hydroxybutyrate, Ethyl 3-hydroxybutyrate
076135-100G
076135-500G
102544-650EA
176.92
USD
102544-650
102664-084
Ethyl-3-hydroxybutyrate ≥95%
Ethyl-3-hydroxybutyrate
Hazard: Irritant
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol |
MDL Number:
MFCD00004545 CAS Number: 5405-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...