3-Hydroxy-2-naphthoic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Hydroxy-naphthalene-2-carboxylic acid, 2-Hydroxy-3-naphthoic acid
076132-10G
076132-100G
102544-636EA
15.38
USD
102544-636
102544-638
3-Hydroxy-2-naphthoic acid ≥95%
3-Hydroxy-2-naphthoic acid
Hazard: Irritant
Formula:
C₁₁H₈O₃ MW: 188.18 g/mol Melting Pt: 218-222°C |
MDL Number:
MFCD00004103 CAS Number: 92-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...