Dehydroacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione, 3-Acetyl-4-hydroxy-6-methyl-2H-pyran-2-one

076129-10G 076129-100G
102544-622EA 36.92 USD
102544-622 102544-624
Dehydroacetic acid ≥95%
Dehydroacetic acid

Hazard: Irritant

Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 333
Melting Pt: 108
Flash Pt: 157
MDL Number: MFCD00066709
CAS Number: 520-45-6
Merck Index: 13,02885

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR