Pyrogallol ≥95%

Supplier: Matrix Scientific

Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene

076126-10G 076126-100G
102544-614EA 36.92 USD
102544-614 102544-616
Pyrogallol ≥95%
Pyrogallol

Hazard: Irritant

Formula: C₆H₆O₃
MDL Number: MFCD00002192
CAS Number: 87-66-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09c5271be2848ebfa6e3614bff56b329a59cb6fe&vid=db25c87d92a56d9ff5d36f04d6bbf59acf236659&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR